2-Chloro-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

14313-25G 14313-5G
76895-426EA 453.01 USD
76895-426 76895-428
2-Chloro-3-methoxybenzoic acid ≥95%
2-Chloro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD07784399
CAS Number: 33234-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR