2-Chloro-4-iodonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-iodopyridine-3-carboxylic acid

14256-25G 14256-5G
76895-390EA 407.39 USD
76895-390 76895-392
2-Chloro-4-iodonicotinic acid ≥97%
2-Chloro-4-iodonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663520
CAS Number: 544671-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR