2-Chloro-3-fluoro-5-iodopyridine ≥97% research grade

Supplier: AOBChem USA

Warning

14213-1G 14213-25G 14213-500MG 14213-5G
76895-378EA 201.58 USD
76895-378 76895-380 76895-382 76895-384
2-Chloro-3-fluoro-5-iodopyridine ≥97% research grade
2-Chloro-3-fluoro-5-iodopyridine
Formula: C₅H₂ClFIN
Storage Temperature: Refrigerator
MDL Number: MFCD09909856
CAS Number: 153034-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR