2-Chloro-3,4-diiodopyridine ≥97%

Supplier: AOBChem USA

Warning

14208-1G 14208-25G 14208-5G
76895-368EA 71.09 USD
76895-368 76895-370 76895-372
2-Chloro-3,4-diiodopyridine ≥97%
2-Chloro-3,4-diiodopyridine
Formula: C₅H₂ClI₂N
Storage Temperature: Refrigerator
MDL Number: MFCD16878974
CAS Number: 153034-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR