2-Fluoro-5-formyl-3-picoline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-5-methylnicotinaldehyde, 2-Fluoro-5-formyl-3-methylpyridine

14193-25G 14193-5G
76895-350EA 488.02 USD
76895-350 76895-352
2-Fluoro-5-formyl-3-picoline ≥97%
2-Fluoro-5-formyl-3-picoline

Precaución: for Research Use Only

Formula: C₇H₆FNO
MW: 139.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659485
CAS Number: 884495-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR