3-Chloro-4-methylbenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-1-methyl-4-(trifluoromethyl)benzene

14165-1G 14165-25G 14165-5G 14165-500MG
76895-332EA 70.02 USD
76895-332 76895-334 76895-336 77648-640
3-Chloro-4-methylbenzotrifluoride ≥97%
3-Chloro-4-methylbenzotrifluoride
Formula: C₈H₆ClF₃
MW: 194.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226657
CAS Number: 74483-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR