3-Formyl-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxynicotinaldehyde, 2-Methoxy-3-pyridinecarboxaldehyde, 2-Methoxy-pyridine-3-carbaldehyde, 2-Methoxypyridine-3-carboxaldehyde

14149-25G 14149-5G
76895-324EA 187.78 USD
76895-324 76895-326
3-Formyl-2-methoxypyridine ≥97%
3-Formyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115112
CAS Number: 71255-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR