3-(3-Chloro-4-fluorophenylcarbamoyl)-4-fluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14089-1G 14089-25G 14089-5G
76895-238EA 64.72 USD
76895-238 76895-240 76895-242
3-(3-Chloro-4-fluorophenylcarbamoyl)-4-fluorophenylboronic acid ≥97% research grade
3-(3-Chloro-4-fluorophenylcarbamoyl)-4-fluorophenylboronic acid
Formula: C₁₃H₉BClF₂NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11856005
CAS Number: 1451393-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR