2-Fluoro-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14031-25G 14031-5G
76895-210EA 183.54 USD
76895-210 76895-212
2-Fluoro-4-formylphenylboronic acid ≥97%
2-Fluoro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MW: 167.9300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367058
CAS Number: 871126-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR