2-Chloro-2,2-difluoro-1-(4-(trifluoromethoxy)phenyl)ethanone ≥97% research grade

Supplier: AOBChem USA

Warning

138436-1G 138436-500MG 138436-5G
76895-164EA 612.14 USD
76895-164 76895-166 76895-168
2-Chloro-2,2-difluoro-1-(4-(trifluoromethoxy)phenyl)ethanone ≥97% research grade
2-Chloro-2,2-difluoro-1-(4-(trifluoromethoxy)phenyl)ethanone
Formula: C₉H₄ClF₅O₂
MW: 274.572 g/mol
Storage Temperature: Ambient
CAS Number: 115446-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR