2-Chloro-2,2-difluoro-1-(3-fluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

138408-1G 138408-25G 138408-500MG 138408-5G
76895-150EA 318.27 USD
76895-150 76895-152 76895-154 76895-156
2-Chloro-2,2-difluoro-1-(3-fluorophenyl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(3-fluorophenyl)ethanone
Formula: C₈H₄ClF₃O
MW: 208.565 g/mol
Storage Temperature: Ambient
CAS Number: 1352222-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR