2-Amino-4-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Amino-4-methylphenylboronic acid, pinacol ester

13818-1G 13818-25G 13818-500MG 13818-5G
76895-134EA 168.69 USD
76895-134 76895-136 76895-138 76895-140
2-Amino-4-methylphenylboronic acid pinacol ester ≥97%
2-Amino-4-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₂
MW: 233.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795679
CAS Number: 863578-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR