2-Cyano-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13706-1G 13706-25G 13706-5G
76895-120EA 114.58 USD
76895-120 76895-122 76895-124
2-Cyano-6-fluorophenylboronic acid ≥97%
2-Cyano-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD09033623
CAS Number: 656235-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR