3-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13642-25G 13642-5G
76895-112EA 375.56 USD
76895-112 76895-114
3-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid ≥97%
3-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027261
CAS Number: 957060-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR