4-Borono-3-fluorothioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-methylthiophenylboronic acid, (2-Fluoro-4-(methylthio)phenyl)boronic acid, 2-Fluoro-4-(methylsulphanyl)benzeneboronic acid

13620-1G 13620-25G 13620-500MG 13620-5G
76895-096EA 289.63 USD
76895-096 76895-098 76895-100 76895-102
4-Borono-3-fluorothioanisole ≥97%
4-Borono-3-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₈BFO₂S
MW: 186.0116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027247
CAS Number: 957060-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR