2-Chloro-3-cyano-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

13561-1G 13561-25G 13561-500MG 13561-5G
76895-084EA 292.81 USD
76895-084 76895-086 76895-088 76895-090
2-Chloro-3-cyano-4-pyridineboronic acid ≥97%
2-Chloro-3-cyano-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09037478
CAS Number: 878194-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR