2,6-Dibromo-3-fluoropyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
13499-25G
13499-5G
76895-072EA
566.53
USD
76895-072
76895-074
2,6-Dibromo-3-fluoropyridine ≥97%
2,6-Dibromo-3-fluoropyridine
Formula:
C₅H₂Br₂FN MW: 254.88 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185513 CAS Number: 41404-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...