3-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(3-Fluorophenylcarbamoyl)benzeneboronic acid

13488-25G 13488-5G
76895-066EA 396.78 USD
76895-066 76895-068
3-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥97%
3-(3-Fluorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027207
CAS Number: 874288-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR