3-Bromo-4-formylbenzonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
13485-1G
13485-25G
13485-5G
76895-056EA
79.57
USD
76895-056
76895-058
76895-060
3-Bromo-4-formylbenzonitrile ≥97%
3-Bromo-4-formylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₄BrNO Storage Temperature: Refrigerator |
MDL Number:
MFCD09261031 CAS Number: 89891-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...