2-Methyl-3-carboxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13413-1G 13413-500MG 13413-5G
76895-030EA 458.31 USD
76895-030 76895-032 76895-034
2-Methyl-3-carboxyphenylboronic acid ≥97%
2-Methyl-3-carboxyphenylboronic acid
Formula: C₈H₉BO₄
MW: 179.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037834
CAS Number: 1314264-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR