2-Chloro-4-(difluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

13300-1G 13300-500MG 13300-5G
76895-010EA 933.6 USD
76895-010 76895-012 76895-014
2-Chloro-4-(difluoromethoxy)benzaldehyde ≥97%
2-Chloro-4-(difluoromethoxy)benzaldehyde
Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32631764
CAS Number: 2222644-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR