2-Hydroxy-3-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

12986-1G 12986-25G 12986-500MG 12986-5G
76894-984EA 287.51 USD
76894-984 76894-986 76894-988 76894-990
2-Hydroxy-3-(trifluoromethoxy)benzoic acid ≥97%
2-Hydroxy-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₄
MW: 222.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD6203613
CAS Number: 773873-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR