2-Acetamido-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Acetamidopyridine-5-boronic acid, pinacol ester, N-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide

12934-1G 12934-25G 12934-5G
76894-962EA 91.24 USD
76894-962 76894-964 76894-966
2-Acetamido-5-pyridineboronic acid pinacol ester ≥97%
2-Acetamido-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
MW: 262.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08690229
CAS Number: 904326-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR