2,6-Dichloro-3-hydroxyisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

129217-1G 129217-25G 129217-500MG 129217-5G
76894-946EA 185.66 USD
76894-946 76894-948 76894-950 76894-952
2,6-Dichloro-3-hydroxyisonicotinic acid ≥97%
2,6-Dichloro-3-hydroxyisonicotinic acid
Formula: C₆H₃Cl₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD03411718
CAS Number: 185422-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR