2,6-Dibromo-4-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

128653-1G 128653-500MG 128653-5G 128653-25G
76894-934EA 876.31 USD
76894-934 76894-936 76894-938 77657-344
2,6-Dibromo-4-methylpyridin-3-ol ≥97%
2,6-Dibromo-4-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34827739
CAS Number: 2852767-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR