2-Fluoro-4-iodonicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-iodo-nicotinic acid

12734-25G 12734-5G
76894-890EA 283.27 USD
76894-890 76894-892
2-Fluoro-4-iodonicotinic acid ≥97% research grade
2-Fluoro-4-iodonicotinic acid
Formula: C₆H₃FINO₂
Storage Temperature: Ambient
MDL Number: MFCD03095310
CAS Number: 884494-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR