3-(1-Methyl-1H-pyrazol-5-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

125014-1G 125014-500MG 125014-5G
76894-868EA 699.14 USD
76894-868 76894-870 76894-872
3-(1-Methyl-1H-pyrazol-5-yl)benzaldehyde ≥97%
3-(1-Methyl-1H-pyrazol-5-yl)benzaldehyde
Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
Storage Temperature: Ambient
CAS Number: 179056-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR