2-Methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-(trifluoromethyl)benzeneboronic acid

12215-25G 12215-5G
76894-824EA 318.27 USD
76894-824 76894-826
2-Methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%
2-Methoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039222
CAS Number: 240139-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR