2-Iodo-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

119961-1G 119961-500MG 119961-5G 119961-25G
76894-730EA 518.79 USD
76894-730 76894-732 76894-734 77653-330
2-Iodo-4-methylbenzaldehyde ≥97%
2-Iodo-4-methylbenzaldehyde
Formula: C₈H₇IO
MW: 246.045 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18397771
CAS Number: 126156-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR