2,6-Dichloropyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

11932-1G 11932-25G 11932-500MG 11932-5G
76894-704EA 144.29 USD
76894-704 76894-706 76894-708 76894-710
2,6-Dichloropyridin-3-ol ≥97%
2,6-Dichloropyridin-3-ol
Formula: C₅H₃Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD11044319
CAS Number: 52764-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR