2,4-Difluoro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,4-Difluoro-3-formylphenyl)boronic acid

11890-25G 11890-5G
76894-690EA 289.63 USD
76894-690 76894-692
2,4-Difluoro-3-formylphenylboronic acid ≥97%
2,4-Difluoro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₃
MW: 185.9200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974116
CAS Number: 870718-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR