2,6-Difluoro-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,6-Difluoropyridin-4-yl)boronic acid, 2,6-Difluoro-4-pyridylboronic acid

11873-1G 11873-25G 11873-500MG 11873-5G
76894-674EA 213.25 USD
76894-674 76894-676 76894-678 76894-680
2,6-Difluoro-4-pyridineboronic acid ≥97%
2,6-Difluoro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BF₂NO₂
MW: 158.8986 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114289
CAS Number: 401816-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR