2,6-Difluoro-4-methoxybenzamide ≥95%
Supplier: AOBChem USA
|
Warning
|
11805-25G
11805-5G
76894-664EA
453.01
USD
76894-664
76894-666
2,6-Difluoro-4-methoxybenzamide ≥95%
2,6-Difluoro-4-methoxybenzamide
Formula:
C₈H₇F₂NO₂ MW: 187.143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094497 CAS Number: 125369-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...