2,6-Difluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

11770-25G 11770-5G
76894-658EA 240.83 USD
76894-658 76894-660
2,6-Difluoro-4-methoxybenzoic acid ≥97%
2,6-Difluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02949623
CAS Number: 123843-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR