2-Fluoro-3-iodo-5-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
11572-25G
11572-5G
76894-622EA
247.19
USD
76894-622
76894-624
2-Fluoro-3-iodo-5-methylpyridine ≥97%
2-Fluoro-3-iodo-5-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅FIN Storage Temperature: Refrigerator |
MDL Number:
MFCD03095303 CAS Number: 153034-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...