2-Bromo-3-fluoro-5-(trifluoromethyl)phenol ≥97% research grade

Supplier: AOBChem USA

Warning

115387-1G 115387-500MG 115387-5G
76894-608EA 744.76 USD
76894-608 76894-610 76894-612
2-Bromo-3-fluoro-5-(trifluoromethyl)phenol ≥97% research grade
2-Bromo-3-fluoro-5-(trifluoromethyl)phenol
Formula: C₇H₃BrF₄O
MW: 258.9957 g/mol
Storage Temperature: Ambient
CAS Number: 1805251-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR