2,5-Dichloro-4-hydroxypyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,5-Dichloropyridin-4-ol
11292-1G
11292-25G
11292-5G
76894-540EA
82.76
USD
76894-540
76894-542
76894-544
2,5-Dichloro-4-hydroxypyridine ≥97%
2,5-Dichloro-4-hydroxypyridine
Precaución: for Research Use Only
Formula:
C₅H₃Cl₂NO MW: 163.989 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01861982 CAS Number: 847664-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...