2-Bromo-4-fluorophenyl ethyl ether ≥97% research grade

Supplier: AOBChem USA

Warning

11123-25G 11123-5G
76894-512EA 506.05 USD
76894-512 76894-514
2-Bromo-4-fluorophenyl ethyl ether ≥97% research grade
2-Bromo-4-fluorophenyl ethyl ether
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD09939735
CAS Number: 326-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR