2-Isopropyl-4-methylanisole ≥97%

Supplier: AOBChem USA

Warning

110894-1G 110894-25G 110894-5G 110894-500MG
76894-504EA 100.79 USD
76894-504 76894-506 76894-508 77652-276
2-Isopropyl-4-methylanisole ≥97%
2-Isopropyl-4-methylanisole

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD18967937
CAS Number: 31574-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR