2-Bromo-3,6-difluorobenzonitrile ≥97% research grade

Supplier: AOBChem USA

Warning

108036-1G 108036-25G 108036-500MG 108036-5G
76894-440EA 231.28 USD
76894-440 76894-442 76894-444 76894-446
2-Bromo-3,6-difluorobenzonitrile ≥97% research grade
2-Bromo-3,6-difluorobenzonitrile
Formula: C₇H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD26687961
CAS Number: 1502090-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR