(2-Methoxy-4-(methoxycarbonyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-4-(methoxycarbonyl)phenylboronic acid

10791-1G 10791-25G 10791-500MG 10791-5G
76894-430EA 201.58 USD
76894-430 76894-432 76894-434 76894-436
(2-Methoxy-4-(methoxycarbonyl)phenyl)boronic acid ≥97% research grade
(2-Methoxy-4-(methoxycarbonyl)phenyl)boronic acid
Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190192
CAS Number: 849758-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR