2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Amino-4-methoxycarbonyl phenylboronic acid Hydrochloride, 2-Amino-4-(methoxycarbonyl)phenylboronic acid hydrochloride, 2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride, 2-Amino-4-methoxycarbonylphenylboronic acid HCl, (2-Amino-4-(methoxycarbonyl)phenyl)boronic acid hydrochloride, 2-Amino-4-(methoxycarbonyl)benzeneboronic acid Hydrochloride

10606-25G 10606-5G
76894-410EA 286.45 USD
76894-410 76894-412
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥95% research grade
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride
Formula: C₈H₁₀BNO₄·HCl
MW: 231.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02258941
CAS Number: 380430-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR