2,4-Dimethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

10538-25G 10538-5G
76894-392EA 244.01 USD
76894-392 76894-394
2,4-Dimethoxyphenylboronic acid pinacol ester ≥97%
2,4-Dimethoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₄
MW: 264.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795688
CAS Number: 214360-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR