2-Fluoro-3-methyl-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-5-methyl-3-pyridylboronic acid, 2-Fluoro-3-methylpyridine-5-boronic acid, (6-Fluoro-5-methylpyridin-3-yl)boronic acid

10495-25G 10495-5G
76894-370EA 215.37 USD
76894-370 76894-372
2-Fluoro-3-methyl-5-pyridineboronic acid ≥97%
2-Fluoro-3-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
MW: 154.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411562
CAS Number: 904326-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR