4-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-ethoxycarbonylphenylboronic acid

10489-1G 10489-25G 10489-5G
76894-360EA 102.91 USD
76894-360 76894-362 76894-364
4-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥97%
4-(Ethoxycarbonyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MW: 211.9827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458482
CAS Number: 874459-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR