2-Fluoro-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

10486-25G 10486-5G
76894-354EA 209 USD
76894-354 76894-356
2-Fluoro-4-pyridineboronic acid pinacol ester ≥97%
2-Fluoro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798253
CAS Number: 458532-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR