2-Methylbenzofuran ≥97%

Supplier: AOBChem USA

Warning

103042-25G 103042-5G
76894-328EA 481.65 USD
76894-328 76894-330
2-Methylbenzofuran ≥97%
2-Methylbenzofuran

Precaución: for Research Use Only

Formula: C₉H₈O
MW: 132.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005850
CAS Number: 4265-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR