2-Bromo-5-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

10234-25G 10234-5G
76894-322EA 403.15 USD
76894-322 76894-324
2-Bromo-5-ethoxybenzaldehyde ≥97%
2-Bromo-5-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD17015551
CAS Number: 43192-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR