2-Carboxy-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Carboxy-5-flourophenylboronic acid

10196-25G 10196-5G
76894-288EA 275.84 USD
76894-288 76894-290
2-Carboxy-5-fluorophenylboronic acid ≥97%
2-Carboxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₄
Storage Temperature: Ambient
MDL Number: MFCD06656266
CAS Number: 874290-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR