2-Benzyloxy-5-bromopyrimidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)-5-bromopyrimidine

10157-25G 10157-5G
76894-266EA 346.92 USD
76894-266 76894-268
2-Benzyloxy-5-bromopyrimidine ≥97%
2-Benzyloxy-5-bromopyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O
MW: 265.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375140
CAS Number: 742058-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR